4,5-Difluor-2-hydroxybenzonitril ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
H26155.03
H26155.03EA
121
EUR
H26155.03
4,5-Difluor-2-hydroxybenzonitril ≥99%
4,5-Difluor-2-hydroxybenzonitril
Purity: 99%
CAS No.: 186590-36-3
Molecular Formula: C7H3F2NO
Formula Weight: 155.10
Melting Point: 161-163°
UN No.: 3439
MDL No.: MFCD08275287
Formel:
C₇H₃F₂NO Molecular Weight: 155,10 g/mol Schmelzpunkt: 161…163 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08275287 CAS-Nummer: 186590-36-3 EINECS: None ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...