3-Hydroxy-5-(trifluormethyl)benzoesäure ≥98%

Lieferant: Thermo Scientific
Achtung

H26157.03
H26157.03EA 218 EUR
H26157.03
3-Hydroxy-5-(trifluormethyl)benzoesäure ≥98%
3-Hydroxy-5-(trifluoromethyl)benzoic acid

Purity: 99% CAS No.: 328-69-8 Molecular Formula: C8H5F3O3 Formula Weight: 206.12 Melting Point: 191-193° MDL No.: MFCD07368783

Formel: C₈H₅F₃O₃
Molecular Weight: 206,12 g/mol
Schmelzpunkt: 191…193 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368783
CAS-Nummer: 328-69-8
EINECS: None

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR