2-Brom-4-nitro(trifluormethoxy)benzol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H26173.03 H26173.06
H26173.03EA 33.9 EUR
H26173.03 H26173.06
2-Brom-4-nitro(trifluormethoxy)benzol ≥98%
2-Brom-4-nitro(trifluormethoxy)benzol

Purity: 98% CAS No.: 200958-40-3 Molecular Formula: C7H3BrF3NO3 Formula Weight: 286.00 Refractive Index: 1.508

Formel: C₇H₃BrF₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04973758
CAS-Nummer: 200958-40-3
EINECS: None

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR