4-Fluor-3-nitrobenzaldehyd ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
H26181.06
H26181.06EA
69
EUR
H26181.06
4-Fluor-3-nitrobenzaldehyd ≥98%
4-Fluoro-3-nitrobenzaldehyde
Purity: 98%
CAS No.: 42564-51-2
Molecular Formula: C7H4FO3
Formula Weight: 169.11
Flash Point:
Formel:
C₇H₄FNO₃ Molecular Weight: 169,11 g/mol Schmelzpunkt: 44…47 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861388 CAS-Nummer: 42564-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...