2,6-Dimethyl-4-nitroanisol ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Methoxy-1,3-dimethyl-5-nitrobenzene

H26204.03 H26204.06
H26204.03EA 30.4 EUR
H26204.03 H26204.06
2,6-Dimethyl-4-nitroanisol ≥99%
2,6-Dimethyl-4-nitroanisol

Purity: 99% CAS No.: 14804-39-8 Molecular Formula: C9H11NO3 Formula Weight: 181.19 Melting Point: 85-92° MDL No.: MFCD00024537

Formel: C₉H₁₁NO₃
Molecular Weight: 181.19 g/mol
Schmelzpunkt: 85…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00024537
CAS-Nummer: 14804-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR