4-Nitro-1H-pyrazole-3-carboxylic acid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Nitro-3-pyrazolecarboxylic acid, 4-Nitropyrazole-3-carboxylic acid

H26224.09
H26224.09EA 124 EUR
H26224.09
4-Nitro-1H-pyrazole-3-carboxylic acid 98%
4-Nitro-1H-pyrazole-3-carboxylic acid

Purity: 98% CAS No.: 5334-40-7 Molecular Formula: C4H3N3O4 Formula Weight: 157.08 Melting Point: ca 195° dec. MDL No.: MFCD00090899

Formel: C₄H₃N₃O₄
Molecular Weight: 157,085 g/mol
Schmelzpunkt: ∼ 195 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00090899
CAS-Nummer: 5334-40-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR