4-(Methylthio)phenol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H26225.09 H26225.18 H26225.30
H26225.09EA 67.8 EUR
H26225.09 H26225.18 H26225.30
4-(Methylthio)phenol ≥98%

Purity: 98% CAS No.: 1073-72-9 Molecular Formula: C7H8OS Formula Weight: 140.21 Odour: Stench Melting Point: 85-87° Boiling Point: 153-156°/20mm MDL No.: MFCD00002351

Formel: C₇H₈OS
Siedepunkt: 153…156 °C (20 mmHg)
Schmelzpunkt: 85…87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002351
CAS-Nummer: 1073-72-9
EINECS: 214-031-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR