Cyclopropylmagnesiumbromid 0.5-0.7 M in THF

Lieferant: Thermo Scientific
Gefahr

H26273.AE
H26273.AEEA 277 EUR
H26273.AE
Cyclopropylmagnesiumbromid 0.5-0.7 M in THF
Cyclopropylmagnesiumbromid

CAS No.: 23719-80-4 Molecular Formula: C3H5BrMg Formula Weight: 145.28 Flash Point: -17°(1°F) UN No.: 2924 MDL No.: MFCD00672094

Formel: C₃H₅BrMg
Molecular Weight: 145,28 g/mol
Dichte: 0,968
Flammpunkt: –17 °C (1 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00672094
CAS-Nummer: 23719-80-4
UN: 2924
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR