4-Cyan-4'-octylbiphenyl ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Cyano-4'-n-octylbiphenyl, 4'-n-Octylbiphenyl-4-carbonitrile, 4'-n-Octyl-4-biphenylcarbonitrile, 8CB

H26318.03
H26318.03EA 143 EUR
H26318.03
4-Cyan-4'-octylbiphenyl ≥99%
4-Cyan-4'-octylbiphenyl

Purity: 99% CAS No.: 52709-84-9 Molecular Formula: C21H25N Formula Weight: 291.43 Flash Point: 110°(230°F) Melting Point: 20-22° MDL No.: MFCD00075146

Formel: C₂₁H₂₅N
Molecular Weight: 291.43 g/mol
Schmelzpunkt: 20…22 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00075146
CAS-Nummer: 52709-84-9
EINECS: 258-120-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR