3-Chlor-5-fluoranilin ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

H26338.03 H26338.06
H26338.03EA 121 EUR
H26338.03 H26338.06
3-Chlor-5-fluoranilin ≥97%
3-Chlor-5-fluoranilin

Purity: 97% CAS No.: 4863-91-6 Molecular Formula: C6H5ClFN Formula Weight: 145.56 Refractive Index: 1.562-1.564 Odour: Characteristic UN No.: 2810 MDL No.: MFCD03407962

Formel: C₆H₅ClFN
Lagertemperatur: Raumtemperatur
MDL: MFCD03407962
CAS-Nummer: 4863-91-6
EINECS: None
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR