3-Chlor-5-fluoranilin ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H26338.03
H26338.06
H26338.03EA
121
EUR
H26338.03
H26338.06
3-Chlor-5-fluoranilin ≥97%
3-Chlor-5-fluoranilin
Purity: 97%
CAS No.: 4863-91-6
Molecular Formula: C6H5ClFN
Formula Weight: 145.56
Refractive Index: 1.562-1.564
Odour: Characteristic
UN No.: 2810
MDL No.: MFCD03407962
Formel:
C₆H₅ClFN Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407962 CAS-Nummer: 4863-91-6 EINECS: None UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...