4-Cyano-5-methylsulphanylthiophene-2-carboxylic acid methyl ester 97%
Lieferant: Thermo Scientific
|
Gefahr
|
H26353.03
H26353.03EA
101
EUR
H26353.03
4-Cyano-5-methylsulphanylthiophene-2-carboxylic acid methyl ester 97%
4-Cyano-5-methylsulphanylthiophene-2-carboxylic acid methyl ester
Purity: 97%
CAS No.: 175202-48-9
Molecular Formula: C8H7NO2S2
Formula Weight: 213.27
Melting Point: 117-119°
UN No.: UN3439
MDL No.: MFCD00173757
Formel:
C₈H₇NO₂S₂ Molecular Weight: 213,28 g/mol Schmelzpunkt: 117…119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173757 CAS-Nummer: 175202-48-9 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...