4-(Ethoxycarbonyl)phenylboronsäure ≥94%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-(Ethoxycarbonyl)benzeneboronic acid, (4-Ethoxycarbonylphenyl)boronic acid

H26356.03 H26356.06
H26356.03EA 29.9 EUR
H26356.03 H26356.06
4-(Ethoxycarbonyl)phenylboronsäure ≥94%
4-(Ethoxycarbonyl)phenylboronsäure

Purity: 94% CAS No.: 4334-88-7 Molecular Formula: C9H11BO4 Formula Weight: 193.99 Melting Point: 137-140° MDL No.: MFCD02179441

Formel: C₉H₁₁BO₄
Schmelzpunkt: 137…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179441
CAS-Nummer: 4334-88-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR