4-Brom-2,3-difluorphenol ≥97%

Lieferant: Thermo Scientific
Gefahr

H26369.06
H26369.06EA 102 EUR
H26369.06
4-Brom-2,3-difluorphenol ≥97%
4-Brom-2,3-difluorphenol

Purity: 97% CAS No.: 144292-32-0 Molecular Formula: C6H3BrF2O Formula Weight: 208.99 MDL No.: MFCD08061907

Formel: C₆H₃BrF₂O
Molecular Weight: 208,99 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061907
CAS-Nummer: 144292-32-0
EINECS: None

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR