5,5'-Dimethyl-2,2'-bipyridin ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5,5'-Dimethyl-2,2'-bipyridine, Abametapir, 6,6'-Bi-3-picoline
H26378.06
H26378.14
H26378.06EA
74.8
EUR
H26378.06
H26378.14
5,5'-Dimethyl-2,2'-bipyridin ≥98%
5,5'-Dimethyl-2,2'-bipyridyl
Purity: 98%
CAS No.: 1762-34-1
Molecular Formula: C12H12N2
Formula Weight: 184.24
Melting Point: 114-117°
Boiling Point: 140°/3mm
MDL No.: MFCD01740554
Formel:
C₁₂H₁₂N₂ Siedepunkt: 140 °C (3 mmHg) Schmelzpunkt: 114…117 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01740554 CAS-Nummer: 1762-34-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...