5,5'-Dimethyl-2,2'-bipyridin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5,5'-Dimethyl-2,2'-bipyridine, Abametapir, 6,6'-Bi-3-picoline

H26378.06 H26378.14
H26378.06EA 74.8 EUR
H26378.06 H26378.14
5,5'-Dimethyl-2,2'-bipyridin ≥98%
5,5'-Dimethyl-2,2'-bipyridyl

Purity: 98% CAS No.: 1762-34-1 Molecular Formula: C12H12N2 Formula Weight: 184.24 Melting Point: 114-117° Boiling Point: 140°/3mm MDL No.: MFCD01740554

Formel: C₁₂H₁₂N₂
Siedepunkt: 140 °C (3 mmHg)
Schmelzpunkt: 114…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01740554
CAS-Nummer: 1762-34-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR