2-Brom-5-(trifluormethoxy)phenol ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

H26389.06 H26389.14
H26389.06EA 93.2 EUR
H26389.06 H26389.14
2-Brom-5-(trifluormethoxy)phenol ≥97%
2-Brom-5-(trifluormethoxy)phenol

Purity: 97% CAS No.: 205371-26-2 Molecular Formula: C7H4BrF3O2 Formula Weight: 257.01 Refractive Index: 1.4815

Formel: C₇H₄BrF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06660162
CAS-Nummer: 205371-26-2
EINECS: None

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR