(4-(Trifluormethyl)phenyl)methansulfonsäurechlorid ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: [4-(Trifluoromethyl)phenyl]methanesulphonyl chloride, 4-Trifluoromethyl-α-toluenesulphonyl chloride

H26395.03
H26395.03EA 151 EUR
H26395.03
(4-(Trifluormethyl)phenyl)methansulfonsäurechlorid ≥97%
(4-(Trifluoromethyl)phenyl)methanesulphonyl chloride

Purity: 97% CAS No.: 163295-75-8 Molecular Formula: C8H6ClF3O2S Formula Weight: 258.65 Melting Point: 103-105° UN No.: 3261 MDL No.: MFCD01631932

Formel: C₈H₆ClF₃O₂S
Molecular Weight: 258.65 g/mol
Schmelzpunkt: 103…105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631932
CAS-Nummer: 163295-75-8
EINECS: None
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR