3-(Brommethyl)pentan ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-(Bromomethyl)pentane
H26445.AA
H26445.AC
H26445.AAEA
22.3
EUR
H26445.AA
H26445.AC
3-(Brommethyl)pentan ≥97%
3-(Brommethyl)pentan
Purity: 97%
CAS No.: 3814-34-4
Molecular Formula: (CH3CH2)2CHCH2Br
Formula Weight: 165.07
Storage Temperature: 2°C to 8°C
Flash Point: 36°(96°F)
Refractive Index: 1.4515
Odour: Sweet
Boiling Point: 145°
UN No.: 1993
MDL No.: MFCD00000219
Formel:
C₆H₁₃Br Siedepunkt: 145 °C Dichte: 1,179 Flammpunkt: 36 °C (96 °F) |
MDL:
MFCD00000219 CAS-Nummer: 3814-34-4 EINECS: 223-302-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...