3,5-Difluor-4-methoxybenzylbromid ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

H26526.03 H26526.06
H26526.03EA 89.2 EUR
H26526.03 H26526.06
3,5-Difluor-4-methoxybenzylbromid ≥97%
3,5-Difluor-4-methoxybenzylbromid

Purity: 97% CAS No.: 706786-42-7 Molecular Formula: C8H7BrF2O Formula Weight: 237.04 Melting Point: 39-42° UN No.: 3261 MDL No.: MFCD04115916

Formel: C₈H₇BrF₂O
Molecular Weight: 237.04 g/mol
Schmelzpunkt: 39…42 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115916
CAS-Nummer: 706786-42-7
UN: 3261
ADR: 8 II

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR