4-Brom-2-nitrophenol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H26529.06 H26529.14
H26529.06EA 25.9 EUR
H26529.06 H26529.14
4-Brom-2-nitrophenol ≥98%
4-Brom-2-nitrophenol

Purity: 98% CAS No.: 7693-52-9 Molecular Formula: C6H4BrNO3 Formula Weight: 218.00 Melting Point: 88-91° MDL No.: MFCD00082540

Formel: C₆H₄BrNO₃
Molecular Weight: 218.01 g/mol
Siedepunkt: 259
Schmelzpunkt: 88…91 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082540
CAS-Nummer: 7693-52-9
EINECS: 231-707-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR