4-Brom-2-nitrophenol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H26529.06
H26529.14
H26529.06EA
25.9
EUR
H26529.06
H26529.14
4-Brom-2-nitrophenol ≥98%
4-Brom-2-nitrophenol
Purity: 98%
CAS No.: 7693-52-9
Molecular Formula: C6H4BrNO3
Formula Weight: 218.00
Melting Point: 88-91°
MDL No.: MFCD00082540
Formel:
C₆H₄BrNO₃ Siedepunkt: 259 °C (1013 hPa) Schmelzpunkt: 88…91 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082540 CAS-Nummer: 7693-52-9 EINECS: 231-707-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...