3-(Difluormethoxy)benzaldehyd ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H26580.03 H26580.06
H26580.03EA 86.7 EUR
H26580.03 H26580.06
3-(Difluormethoxy)benzaldehyd ≥97%
3-(Difluormethoxy)benzaldehyd

Purity: 97% CAS No.: 85684-61-3 Molecular Formula: C8H6F2O2 Formula Weight: 172.13 MDL No.: MFCD00236222

Formel: C₈H₆F₂O₂
Molecular Weight: 172.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236222
CAS-Nummer: 85684-61-3
EINECS: None

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR