(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (AOP) ) ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: ((3H-[1,2,3]Triazolo[4,5-b]pyridin-3-yl)oxy)tris(dimethylamino)phosphonium hexafluorophosphate(V), 7-Aza-1H-benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, (7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium Hexafluorophosphate, (7-Azabenzotriazol-1-yl)-N-oxy-tris(dimethyamino)phosphonium hexafluorosphate, AOP, Tris(dimethylamino)(3H-1,2,3-triazolo[4,5-b]pyridin-3-yloxy)phosphonium Hexafluorophosphate

H26609.06
H26609.06EA 111 EUR
H26609.06
(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (AOP) ) ≥98%
(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (AOP) )

Purity: 98+% CAS No.: 156311-85-2 Molecular Formula: C11H21F6N7OP2 Formula Weight: 443.27 Storage Temperature: 2°C to 8°C Melting Point: ca 180° dec. MDL No.: MFCD09263289

Formel: C₁₂H₂₂F₆N₆OP₂
Molecular Weight: 443,27 g/mol
Schmelzpunkt: ∼ 180 °C (Zers.)
MDL: MFCD09263289
CAS-Nummer: 156311-85-2
EINECS: None

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR