3-(Ethoxycarbonyl)phenylboronsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-(Ethoxycarbonyl)benzeneboronic acid, 3-Ethoxycarbonylphenylboronic acid

H26615.03 H26615.06
H26615.03EA 65.2 EUR
H26615.03 H26615.06
3-(Ethoxycarbonyl)phenylboronsäure ≥97%
3-(Ethoxycarbonyl)phenylboronsäure

Purity: 97% CAS No.: 4334-87-6 Molecular Formula: C9H11BO4 Formula Weight: 193.99 Melting Point: 135-139° MDL No.: MFCD02179444

Formel: C₉H₁₁BO₄
Molecular Weight: 193.99 g/mol
Schmelzpunkt: 135…139 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179444
CAS-Nummer: 4334-87-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR