1,8-Dibenzyl-1,4,8,11-tetraazacyclotetradecane
Lieferant: Thermo Scientific
|
Achtung
|
H26643.MD
H26643.MDEA
81.5
EUR
H26643.MD
1,8-Dibenzyl-1,4,8,11-tetraazacyclotetradecane
1,8-Dibenzyl-1,4,8,11-tetraazacyclotetradecane
CAS No.: 214078-93-0
Molecular Formula: C24H36N4
Formula Weight: 380.58
Melting Point: 67-68°
MDL No.: MFCD09263312
Formel:
C₂₄H₃₆N₄ Molecular Weight: 380,58 g/mol Schmelzpunkt: 67…68 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09263312 CAS-Nummer: 214078-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...