3-(Benzylamino)-1-propanol 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(Benzylamino)propan-1-ol, N-(3-Hydroxypropyl)benzylamine, 3-(Benzylamino)-1-propanol
H26664.03
H26664.03EA
212
EUR
H26664.03
3-(Benzylamino)-1-propanol 97%
3-(Benzylamino)-1-propanol
Purity: 97%
CAS No.: 4720-29-0
Molecular Formula: C6H5CH2NH(CH2)3OH
Formula Weight: 165.24
MDL No.: MFCD00020622
Formel:
C₁₀H₁₅NO Molecular Weight: 165,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00020622 CAS-Nummer: 4720-29-0 EINECS: 225-210-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...