2-Acetylquinoxaline 97%

Lieferant: Thermo Fisher Scientific
Achtung

H26680.03
H26680.03EA 173 EUR
H26680.03
2-Acetylquinoxaline 97%
2-Acetylquinoxaline

Purity: 97% CAS No.: 25594-62-1 Molecular Formula: C10H8N2O Formula Weight: 172.19 Melting Point: 76-79° MDL No.: MFCD07808559

Formel: C₁₀H₈N₂O
Molecular Weight: 172.19 g/mol
Schmelzpunkt: 76…79 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07808559
CAS-Nummer: 25594-62-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR