Agar Cell culture_plant_tested

Lieferant: Thermo Fisher Scientific

Synonyme: Agar Agar

H26724.0E H26724.22 H26724.36
H26724.0EEA 1132 EUR
H26724.0E H26724.22 H26724.36
Agar Cell culture_plant_tested
Agar

CAS No.: 9002-18-0 Odour: Odourless Merck Reference: 14,184 MDL No.: MFCD00081288

Schmelzpunkt: 90
Lagertemperatur: Raumtemperatur
MDL: MFCD00081288
CAS-Nummer: 9002-18-0
EINECS: 232-658-1
Merck Index: 14,184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR