Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7-triacetate

Lieferant: Thermo Scientific
Achtung

Synonyme: 1,4,7,10-Tetraazacyclododecane-1,4,7-triacetic Acid Tri-tert-butyl Ester, Tri-tert-butyl 2,2',2''-(1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate

H26736.MD
H26736.MDEA 225 EUR
H26736.MD
Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7-triacetate
Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7-triacetate

CAS No.: 122555-91-3 Molecular Formula: C26H50N4O6 Formula Weight: 514.71 MDL No.: MFCD05865261

Formel: C₂₆H₅₀N₄O₆
Molecular Weight: 514,71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05865261
CAS-Nummer: 122555-91-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR