2-(Ethoxycarbonyl)ethylzinc bromide 0.5 M in THF

Lieferant: Thermo Fisher Scientific
Gefahr

H26739.AD
H26739.ADEA 207 EUR
H26739.AD
2-(Ethoxycarbonyl)ethylzinc bromide 0.5 M in THF
2-(Ethoxycarbonyl)ethylzinc bromide

CAS No.: 193065-68-8 Molecular Formula: BrZnCH2CH2COOCH2CH3 Formula Weight: 246.42 Storage Temperature: 2°C to 8°C Flash Point: -17°(1°F) UN No.: UN3399 MDL No.: MFCD00671976

Formel: BrZnCH₂CH₂COOCH₂CH₃
Molecular Weight: 246.42 g/mol
Dichte: 0.972
Flammpunkt: –17 °C (1 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00671976
CAS-Nummer: 193065-68-8
UN: 3399
ADR: 4.3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR