1,4,8-Tri-boc-1,4,8,11-tetraazacyclotetradecane

Lieferant: Thermo Scientific
Achtung

Synonyme: Tri-tert-butyl 1,4,8,11-tetraazacyclotetradecane-1,4,8-tricarboxylate

H26741.MD
H26741.MDEA 152 EUR
H26741.MD
1,4,8-Tri-boc-1,4,8,11-tetraazacyclotetradecane
1,4,8-Tri-boc-1,4,8,11-tetraazacyclotetradecane

CAS No.: 170161-27-0 Molecular Formula: C25H48N4O6 Formula Weight: 500.68 Melting Point: 48-54° MDL No.: MFCD09263315

Formel: C₂₅H₄₈N₄O₆
Molecular Weight: 500,68 g/mol
Schmelzpunkt: 48…54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09263315
CAS-Nummer: 170161-27-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR