3-(4-Bromphenyl)propionsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H26785.06
H26785.06EA
47.1
EUR
H26785.06
3-(4-Bromphenyl)propionsäure ≥98%
3-(4-Bromphenyl)propionsäure
Purity: 98%
CAS No.: 1643-30-7
Molecular Formula: C9H9BrO2
Formula Weight: 229.07
Melting Point: 133-136°
MDL No.: MFCD01310793
Formel:
C₉H₉BrO₂ Schmelzpunkt: 133…136 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01310793 CAS-Nummer: 1643-30-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...