4-Fluor-2-methoxybenzylamin 97%

Lieferant: Thermo Fisher Scientific
Gefahr

H26796.03 H26796.06
H26796.03EA 85.6 EUR
H26796.03 H26796.06
4-Fluor-2-methoxybenzylamin 97%
4-Fluor-2-methoxybenzylamin

Purity: 97% CAS No.: 870563-60-3 Molecular Formula: C8H10FNO Formula Weight: 155.17 Refractive Index: 1.5240

Formel: C₈H₁₀FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD04116361
CAS-Nummer: 870563-60-3
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR