4-Carboxyphenylboronsäurepinakolester ≥97%

Lieferant: Thermo Scientific

Synonyme: 4-Carboxybenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

H26825.06
H26825.06EA 105 EUR
H26825.06
4-Carboxyphenylboronsäurepinakolester ≥97%
4-Carboxyphenylboronsäurepinakolester

Purity: 97% CAS No.: 180516-87-4 Molecular Formula: C13H17BO4 Formula Weight: 248.08 Melting Point: 227-231° MDL No.: MFCD01863710

Formel: C₁₃H₁₇BO₄
Molecular Weight: 248,08 g/mol
Schmelzpunkt: 227…231 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01863710
CAS-Nummer: 180516-87-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR