7-Fluorisatin ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 7-Fluorindolin-2,3-dion, 7-Fluor-2,3-indolindion, 7-Fluor-2,3-dioxoindolin, 7-Fluor-1H-indol-2,3-dion

H26827.03 H26827.06 H26827.14
H26827.03EA 28.3 EUR
H26827.03 H26827.06 H26827.14
7-Fluorisatin ≥97%
7-Fluorisatin

Purity: 97% CAS No.: 317-20-4 Molecular Formula: C8H4FNO2 Formula Weight: 165.12 Melting Point: 193-195° MDL No.: MFCD01569508

Formel: C₈H₄FNO₂
Molecular Weight: 165.12 g/mol
Schmelzpunkt: 193…195 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01569508
CAS-Nummer: 317-20-4
EINECS: None

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR