2-Formyl-6-picolin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6-Methylpyridine-2-carboxaldehyde, 6-Methyl-2-pyridinecarboxaldehyde

H26841.14
H26841.14EA 588 EUR
H26841.14
2-Formyl-6-picolin ≥98%
2-Formyl-6-picolin

Purity: 98% CAS No.: 1122-72-1 Molecular Formula: C7H7NO Formula Weight: 121.14 Flash Point: 68°(155°F) Refractive Index: 1.527 Melting Point: 29-33° Boiling Point: 105-108°/12mm Beilstein Reference: 107476 MDL No.: MFCD00006291

Formel: C₇H₇NO
Molecular Weight: 121.14 g/mol
Siedepunkt: 105…108 °C (12 mmHg)
Schmelzpunkt: 29…33 °C
Flammpunkt: 68 °C (155 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006291
CAS-Nummer: 1122-72-1
EINECS: 214-359-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR