1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene ≥99%

Lieferant: Thermo Scientific

Synonyme: 1-Ethyl-4-(p-tolylethynyl)benzene, 1-Ethyl-4-[(p-tolyl)ethynyl]benzene, 1-Ethyl-4-[(4-methylphenyl)ethynyl]benzene, 1-(4-Ethylphenyl)-2-(p-tolyl)acetylene

H26882.03
H26882.03EA 24.8 EUR
H26882.03
1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene ≥99%
1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene

Purity: 99+% CAS No.: 22692-80-4 Molecular Formula: C17H16 Formula Weight: 220.31 Melting Point: 71-74° Boiling Point: 135-136°/1mm MDL No.: MFCD04038793

Formel: C₁₇H₁₆
Molecular Weight: 220,315 g/mol
Siedepunkt: 135…136 °C (1 mmHg)
Schmelzpunkt: 71…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04038793
CAS-Nummer: 22692-80-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR