1,4-Difluor-2-iodbenzol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,5-Difluoroiodobenzene
H26890.06
H26890.06EA
87.7
EUR
H26890.06
1,4-Difluor-2-iodbenzol ≥97%
1,4-Difluoro-2-iodobenzene
Purity: 97%
CAS No.: 2265-92-1
Molecular Formula: C6H3F2I
Formula Weight: 239.99
Flash Point: 85°(185°F)
Refractive Index: 1.5630
Boiling Point: 178-179°
MDL No.: MFCD03094151
Formel:
C₆H₃F₂I Molecular Weight: 239,99 g/mol Siedepunkt: 178…179 °C Dichte: 1,998 Flammpunkt: 85 °C (185 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094151 CAS-Nummer: 2265-92-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...