[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H26897.03
H26897.03EA
61.8
EUR
H26897.03
[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride
[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride
CAS No.: 59831-02-6
Molecular Formula: C27H26Cl2P2Pd
Formula Weight: 589.77
MDL No.: MFCD03844773
Formel:
C₂₇H₂₆Cl₂P₂Pd Lagertemperatur: Raumtemperatur |
MDL:
MFCD03844773 CAS-Nummer: 59831-02-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...