2-Chlor-4-methoxyphenylboronsäure 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Chloro-4-methoxybenzeneboronic acid

H26917.06
H26917.06EA 269 EUR
H26917.06
2-Chlor-4-methoxyphenylboronsäure 95%
2-Chlor-4-methoxyphenylboronsäure

Purity: 95% CAS No.: 219735-99-6 Molecular Formula: C7H8BClO3 Formula Weight: 186.40 Melting Point: ca 202° MDL No.: MFCD03411935

Formel: C₇H₈BClO₃
Schmelzpunkt: ∼ 202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03411935
CAS-Nummer: 219735-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR