(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid 97%

Lieferant: Thermo Scientific

Synonyme: 3-Amino-N-Boc-L-alanine, Nalpha-Boc-2,3-diaminopropionic Acid, Boc-Dpr-OH, Boc-Dap-OH, 3-Amino-N-(tert-butoxycarbonyl)-L-alanine

H26919.03
H26919.03EA 60.1 EUR
H26919.03
(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid 97%
(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid

Purity: 97% CAS No.: 73259-81-1 Molecular Formula: C8H16N2O4 Formula Weight: 204.23 Melting Point: ca 210° dec. Optical Rotation: -14° (c=1 in water) MDL No.: MFCD00236843

Formel: C₈H₁₆N₂O₄
Molecular Weight: 204,23 g/mol
Schmelzpunkt: ∼ 210 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00236843
CAS-Nummer: 73259-81-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR