4'-Acetamido-2'-methylacetophenon ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
N-(4-Acetyl-3-methylphenyl)acetamide
H26933.14
H26933.06
H26933.14EA
114
EUR
H26933.14
H26933.06
4'-Acetamido-2'-methylacetophenon ≥97%
4'-Acetamido-2'-methylacetophenon
Purity: 97%
CAS No.: 34956-31-5
Molecular Formula: C11H13NO2
Formula Weight: 191.23
Melting Point: 137-140°
MDL No.: MFCD02258876
Formel:
C₁₁H₁₃NO₂ Molecular Weight: 191.23 g/mol Schmelzpunkt: 137…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02258876 CAS-Nummer: 34956-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...