4'-Acetamido-2'-methylacetophenon ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-(4-Acetyl-3-methylphenyl)acetamide

H26933.14 H26933.06
H26933.14EA 114 EUR
H26933.14 H26933.06
4'-Acetamido-2'-methylacetophenon ≥97%
4'-Acetamido-2'-methylacetophenon

Purity: 97% CAS No.: 34956-31-5 Molecular Formula: C11H13NO2 Formula Weight: 191.23 Melting Point: 137-140° MDL No.: MFCD02258876

Formel: C₁₁H₁₃NO₂
Molecular Weight: 191.23 g/mol
Schmelzpunkt: 137…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02258876
CAS-Nummer: 34956-31-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR