1-(2-Tetrahydropyranyl)-1H-pyrazol-5-boronsäurepinakolester 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
1-(Tetrahydropyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole-5-boronic acid Pinacol ester, 1-(Tetrahydropyran-2-yl)-1H-pyrazole-5-boronic acid pinacol ester, 1-(Tetrahydro-2H-pyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
H27036.03
H27036.03EA
111
EUR
H27036.03
1-(2-Tetrahydropyranyl)-1H-pyrazol-5-boronsäurepinakolester 95%
1-(2-Tetrahydropyranyl)-1H-pyrazol-5-boronsäurepinakolester
Purity: 95%
CAS No.: 903550-26-5
Molecular Formula: C14H23BN2O3
Formula Weight: 278.16
MDL No.: MFCD09037501
Formel:
C₁₄H₂₃BN₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09037501 CAS-Nummer: 903550-26-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...