1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazol-5-boronsäurepinakolester 95%

Lieferant: Thermo Fisher Scientific

Synonyme: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-pyrazole

H27043.03
H27043.03EA 197 EUR
H27043.03
1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazol-5-boronsäurepinakolester 95%
1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazol-5-boronsäurepinakolester

Purity: 95% CAS No.: 903550-12-9 Molecular Formula: C15H29BN2O3Si Formula Weight: 324.31 MDL No.: MFCD09037502

Formel: C₁₅H₂₉BN₂O₃Si
Lagertemperatur: Raumtemperatur
MDL: MFCD09037502
CAS-Nummer: 903550-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR