(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (2S)-1-[(tert-butoxy)carbonyl]-4,4-difluoropyrrolidine-2-carboxylic acid, (S)-1-(tert-Butoxycarbonyl)-4,4-difluoropyrrolidine-2-carboxylic acid, N-Boc-4,4-difluoro-L-proline, (S)-1-Boc-4,4-difluoro-2-pyrrolidinecarboxylic Acid, N-(tert-Butoxycarbonyl)-4,4-difluoro-L-proline

H27052.03
H27052.03EA 145 EUR
H27052.03
(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid ≥97%
(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid

Purity: 97+% CAS No.: 203866-15-3 Molecular Formula: C10H15F2NO4 Formula Weight: 251.24 Melting Point: 120-124° MDL No.: MFCD03094917

Formel: C₁₀H₁₅F₂NO₄
Molecular Weight: 251,23 g/mol
Schmelzpunkt: 120…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094917
CAS-Nummer: 203866-15-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR