2-Amino-5-chlor-3-iodpyridin 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Chlor-3-iodpyridin-2-amin, 5-Chlor-3-iod-2-pyridylamin

H27053.03
H27053.03EA 74.5 EUR
H27053.03
2-Amino-5-chlor-3-iodpyridin 95%
2-Amino-5-chlor-3-iodpyridin

Purity: 95% CAS No.: 211308-81-5 Molecular Formula: C5H4ClIN2 Formula Weight: 254.46 Melting Point: 95-98° MDL No.: MFCD07644578

Formel: C₅H₄ClIN₂
Molecular Weight: 254,46 g/mol
Schmelzpunkt: 95…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07644578
CAS-Nummer: 211308-81-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR