2-(Diphenylphosphino)ethylamine 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H27070.03
H27070.03EA
191
EUR
H27070.03
2-(Diphenylphosphino)ethylamine 95%
2-(Diphenylphosphino)ethylamine
Purity: 95%
CAS No.: 4848-43-5
Molecular Formula: (C6H5)2PCH2CH2NH2
Formula Weight: 229.27
UN No.: UN2735
MDL No.: MFCD00233831
Formel:
(C₆H₅)₂PCH₂CH₂NH₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00233831 CAS-Nummer: 4848-43-5 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...