3,5-Dimethyladamantane-1-carboxylic acid 97%
Lieferant: Thermo Scientific
|
Achtung
|
H27073.03
H27073.03EA
53.4
EUR
H27073.03
3,5-Dimethyladamantane-1-carboxylic acid 97%
3,5-Dimethyladamantane-1-carboxylic acid
Purity: 97%
CAS No.: 14670-94-1
Molecular Formula: C13H20O2
Formula Weight: 208.30
Melting Point: 107-109°
MDL No.: MFCD00188067
Formel:
C₁₃H₂₀O₂ Molecular Weight: 208,3 g/mol Schmelzpunkt: 107…109 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00188067 CAS-Nummer: 14670-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...